Learn More
R&D Systems™ Human GPR39 (NP_001499) VersaClone cDNA
G Protein-coupled Receptor 39
Brand: R&D Systems™ RDC0245
This item is not returnable.
View return policy
Description
The Human GPR39 (NP_001499) VersaClone cDNA clone contains the complete ORF for GPR39, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The GPR39 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
hGPR39 | |
10 μg at 0.2 μg/μL | |
1375bp | |
Human |
NP_001499 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
Product Content Correction
Product Title
For Reasearch Use Only; Not for Use in Humans