Learn More
R&D Systems™ Human Pluripotent Stem Cell Marker Antibody Panel Plus
Designed for users who are interested in characterizing the status of undifferentiated human ES cells
Brand:  R&D Systems™ SC009
665.21 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18441720
  • 845.00€ / 1 set
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
Includes: Mouse anti-CD9, Mouse anti-E-Cadherin, Goat anti-Nanog, Goat anti-Oct-3/4, Mouse anti-PODXL (GCTM antigen), Mouse anti-SOX2, Mouse anti-SSEA-1, Mouse anti-SSEA-4
Description
Description
Eight markers for increased confidence in pluripotency status: anti-CD9, anti-E-Cadherin, anti-Nanog, anti-Oct-3/4, anti-PODXL (GCTM antigen), anti-SOX2, anti-SSEA-1, and anti-SSEA-4
Verifies the pluripotency of your starting stem cell population
Cost-e
Specifications
Specifications
1 Kit
Store the unopened product at 2°C to 8°C. Do not use past expiration date.