Learn More
R&D Systems™ Mouse CCR5 (AAC52454) VersaClone cDNA
Chemokine (C-C motif) Receptor 5
Brand: R&D Systems™ RDC0011
This item is not returnable.
View return policy
Description
The Mouse CCR5 (AAC52454) VersaClone cDNA clone contains the complete ORF for CCR5, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CCR5 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
mCCR5 | |
10 μg at 0.2 μg/μL | |
1078bp | |
Mouse |
AAC52454 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
Product Content Correction
Product Title
For Reasearch Use Only; Not for Use in Humans