Learn More
R&D Systems™ Mouse Mesenchymal Stem Cell Marker Antibody Panel
For the verification of mesenchymal stem/stromal cell identity by flow cytometry
Brand:  R&D Systems™ SC018
760.24 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18491890
  • 966.00€ / 1 set
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
Includes: 25μg each of antibodies to Sca-1, CD106, CD105, CD73, CD29, CD44, CD11b, and CD45
Description
Description
Researchers use different techniques to isolate, culture, and differentiate human mesenchymal stem/stromal cells (MSCs). Variations in experimental approaches as well as differences in the MSC starting population may account for experimental variability a
Cost-effective panel of premium quality antibodies for the positive and negative identification of MSCs
Uses 9 markers to increase confidence in MSC identification
Enables verification of MSC identity in less than 2 hours with use of fluorochrome-conjug
Specifications
Specifications
1 Kit
Store the unopened product at 2°C to 8°C. Do not use past expiration date.