Learn More
R&D Systems™ Mouse TLR6 (NP_035734) VersaClone cDNA
Toll-like Receptor 6
Brand:  R&D Systems™ RDC0797
Product Code. 18461730
  • 747.00€ / 10µg
Estimated Shipment: 30-01-2025
Description
The Mouse TLR6 (NP_035734) VersaClone cDNA clone contains the complete ORF for TLR6, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The TLR6 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
mTLR6
10 μg at 0.2 μg/μL
2433bp
Mouse
NP_035734
cDNA is provided in 10mM Tris-Cl, pH 8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
For Reasearch Use Only; Not for Use in Humans