Learn More
R&D Systems™ StemXVivo Methylcellulose Concentrate
For the growth and differentiation of human, mouse and rat hematopoietic stem cells using Iscove'ss Modified Dulbecco's Media or any other basal media
Brand:  R&D Systems™ HSC011
Product Code. 18472630
  • 194.00€ / 50mL
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Methylcellulose Concentrate, semisolid medium
50 mL