Learn More
R&D Systems™ Human SLC15A3 (NP_057666) VersaClone cDNA
Solute Carrier Family 15 (Oligopeptide Transporter), Member 3
Brand:  R&D Systems™ RDC1070
Product Code. 18703339
  • 322.00€ / 10µg
Estimated Shipment: 28-11-2024
Description
The Human SLC15A3 (NP_057666) VersaClone cDNA clone contains the complete ORF for SLC15A3, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The SLC15A3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifications
Specifications
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Special Offers
For Reasearch Use Only; Not for Use in Humans