missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ RAB9A 293T Cell Transient Overexpression Lysate (Denatured) (T02)

Human overexpression lysate

Brand:  Abnova™ H00009367-T02.100uL


Product Code. 16100486

  • 215.00€ / 100µL
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

293T
RAB9A 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 22.8kDa
Transient overexpression cell lysate was tested with anti-RAB9A antibody (Mfr. No. H00009367-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
NM_004251
pCMV-RAB9A full-length
100 μL
Human
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 100UL RAB9A 293T Cell Transient Overexpression Lys

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contains : Mercaptoethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.