missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Donkey IgG (H+L) Secondary Antibody, AP, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen PA130343
This item is not returnable.
View return policy
Description
PA1-30343 detects Donkey IgG from donkey samples.
Anti-Donkey secondary antibodies are affinity-purified antibodies with well-characterized specificity for equine (Donkey) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Donkey IgG (H+L) | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Rabbit | |
IgG fraction | |
RUO | |
Donkey | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
0.9 mg/mL | |
0.05M tris chloride with 0.001M MgCl2, 0.001M ZnCl2, 0.15M NaCl, 10mg/mL BSA, 50% glycerol and 0.1% sodium azide; pH 8 | |
donkey IgG whole molecule | |
1 mg | |
Secondary | |
4° C, do not freeze | |
IgG |