missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Donkey IgG whole molecule,Polyclonal Secondary Antibody, Abnova™
Brand: Abnova PAB9199.2mL
This item is not returnable.
View return policy
Specifications
Rabbit anti-Donkey IgG whole molecule | |
Polyclonal | |
Rabbit anti-donkey IgG recognizes donkey IgG whole molecule. This secondary antibody was purified using delipidation and defibrination. | |
Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
Donkey IgG whole molecule | |
2 mL | |
Donkey | |
Lyophilized |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Rabbit | |
This product was prepared from monospecific antiserum by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against donkey IgG and donkey serum. | |
Secondary | |
Store at 4°C on dry atmosphere. After reconstitution with 2mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Rabbit anti-Donkey IgG whole molecule,Polyclonal Secondary Antibody, Abnova™ > 2mL; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction