Learn More
Rabbit anti-Goat IgG Fc Secondary Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-74853APC
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Goat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay
Specifications
IgG Fc | |
Polyclonal | |
PBS | |
Purified Goat IgG Fc fragment | |
1 mL | |
Secondary | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay (CIA) | |
APC | |
Rabbit | |
Affinity purified | |
RUO | |
Goat | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.