Learn More
Rabbit anti-Goat IgG Fc Secondary Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-73851
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Goat samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Fluorophore-linked immunosorbent assay
Specifications
IgG Fc | |
Polyclonal | |
FITC | |
Lyophilized from 0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free | |
Goat IgG F(c) fragment | |
20 mg | |
Adaptive Immunity, Immunology | |
Reconstitute with 2.0 mL deionized water (or equivalent). | |
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-Linked Immunosorbent Assay | |
LYOPH | |
Flow Cytometry 1:500 - 1:2500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
Rabbit | |
Multi-step | |
RUO | |
Secondary | |
Goat | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.