missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Goat IgG (H+L) Secondary Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-72707APC

Product Code. 30226938

  • 343.40€ / 0.50mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Fluorophore-linked immunosorbent assay
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgG (H+L)
Polyclonal
PBS
Goat IgG whole molecule
0.5 mL
Adaptive Immunity, Immunology
Goat
Purified
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Fluorophore-Linked Immunosorbent Assay
APC
Rabbit
Immunogen affinity purified
RUO
Secondary
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rabbit anti-Goat IgG (H+L) Secondary Antibody, Allophycocyanin, Novus Biologicals™ > 0.5 mL; APC

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.