missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Guinea Pig IgG Fc Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP1-72634
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Guinea Pig samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
IgG Fc | |
Polyclonal | |
Western Blot 1:500 - 1:2500, ELISA 1:5000 - 1:15000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200, Dot Blot | |
Guinea Pig IgG F(c) fragment | |
1 mg | |
Secondary | |
Guinea Pig | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot | |
Alkaline Phosphatase (AP) | |
Rabbit | |
Affinity Purified | |
RUO | |
No reaction was observed against Guinea Pig IgG F(ab'). | |
Store at 4C. Do not freeze. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Rabbit anti-Guinea Pig IgG Fc Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™ > 1mg; AP
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction