missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG (H+L) Secondary Antibody, Biotin (Pre-adsorbed), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NB120-7074
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
IgG (H+L) | |
Polyclonal | |
Biotin | |
Rabbit | |
Multi-step | |
RUO | |
Secondary | |
Mouse | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
2.0 mg/mL | |
Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000 | |
Mouse IgG whole molecule | |
1 mg | |
Adaptive Immunity, Immunology | |
This antibody was pre-adsorbed against Human Serum Proteins. No reaction was observed against Human Serum Proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Rabbit anti-Mouse IgG (H+L) Secondary Antibody, Biotin (Pre-adsorbed), Novus Biologicals™ > 1mg; Biotin
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction