missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Rat IgG F(c) fragment, HRP, Polyclonal, Secondary Antibody, Abnova™
Brand: Abnova PAB10882.1500ug
This item is not returnable.
View return policy
Specifications
Rabbit anti-Rat IgG F(c) fragment | |
Polyclonal | |
Rabbit anti-rat IgG recognizes rat IgG F(c) fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Peroxidase. | |
Lyophilized from 10mM potassium phosphate, 75mM NaCl, 50% (v/v) glycerol, pH 7.2 (0.01% (w/v) gentamicin sulfate (Do NOT add sodium azide!), 10mg/mL BSA (Immunoglobulin and Protease free)) | |
Rat IgG F(c) fragment | |
1500 ÎĽg | |
Rat | |
Lyophilized |
Dot Blot, ELISA, Immunohistochemistry, Western Blot | |
HRP | |
The optimal working dilution should be determined by the end user. | |
Rabbit | |
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
Secondary | |
Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Rabbit anti-Rat IgG F(c) fragment, HRP, Polyclonal, Secondary Antibody, Abnova™ > 1500μg; HRP
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction