Learn More
Rabbit IgG Franction to Hamster IgG, Secondary Antibody, MP Biomedicals™
Rabbit anti- Hamster IgG (Whole Molecule) Secondary Antibody
Brand:  MP Biomedicals 0855398
Product Code. 15543467
  • 190.00€ / 2mL
Description
  • Product is the lyophilized powder of rabbit IgG fraction to hamster IgG (whole molecule) and buffer salts.
  • The IgG fraction is prepared from the specific rabbit antiserum by delipidation, ammonium sulfate fractionation and anion exchange chromatography
  • This product is suitable for use as a precipitating antibody. It gives sharp precipitin lines in immunoelectrophoresis and immunodiffusion techniques.
  • This product is suitable for conjugation with enzymes, radiolabels, fluorochromes or other markers, and for attachment to solid supports for use as an immunoadsorbant.
  • This product is suitable for use as a primary reagent in enzyme immunoassays, fluorescein immunoassays and blot, cell or tissue immunostaining.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunohistochemistry, ELISA
Product is the lyophilized powder of rabbit IgG fraction to hamster IgG (whole molecule) and buffer salts
ammonium sulfate fractionation and anion exchange chromatography
RUO
Reconstitute product with 5.0mL of deionized or distilled water.gentle swirling may be used to speed rehydration. Avoid vigorous shaking of the reconstituted material.
Store lyophilized product at 2-8°C. For long term storage, aliquot and store reconstituted product frozen at -20°C or lower.
Unconjugated
Rabbit
2 mL
Secondary
Hamster
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Rabbit IgG Franction to Hamster IgG, Secondary Antibody, MP Biomedicals™ > 2mL, Unlabeled