Learn More
Rabbit Pure-Blot anti-Mouse IgG Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-23974
Product Code. 18650955
  • 345.00€ / 1mg
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
IgG Polyclonal antibody specifically detects IgG in Mouse samples. It is validated for ELISA
Specifications
Specifications
IgG
Polyclonal
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.1 mg/mL Bovine Serum Albumin (BSA) - IgG and Protease free, 50% (v/v) Glycerol
Rabbit
Affinity purified
RUO
Secondary
Mouse
Purified
ELISA
Unconjugated
Immunoglobulin G, ImmunoglobulinG
highly purified mouse IgG gamma 1, gamma 2a, gamma 2b and gamma 3 proteins
1 mg
Adaptive Immunity, Epitope Tags, Immunology
3500
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.