missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals rac-Norgestrel

Product Code. 17201172
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6533-00-2
Molecular Formula C21H28O2

Specifications

Quantity 1 g
Melting Point 205.0°C to 207.0°C
Formula Weight 312.45
InChI Key WWYNJERNGUHSAO-XUDSTZEESA-N
SMILES CC[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C
IUPAC Name (1R,3aS,3bR,9aR,9bS,11aS)-11a-ethyl-1-ethynyl-1-hydroxy-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Molecular Weight (g/mol) 312.45
Chemical Name or Material rac-Norgestrel

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'