missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Rack

Brand:  Tenak™ TE21352

Product Code. 11597214

  • 88.75€ / Each
Estimated Shipment: 05-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

freezer, for 10 boxes, 32mm high, stainless steel, with locking rod, Tenak
Specifications

Specifications

Stainless Steel
Locking rod to secure cryoboxes
140 x 140 x 359 mm
359 mm
Cryoboxes (10, 32 mm H)
All freezer models
1 x 10
140 mm
140 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.