missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ Ramboldi™ High Temperature Autoclave Bags

Ramboldi™ High Temperature Autoclave Bags are printed with blue hazard warnings labels for contaminated waste disposal.

Brand:  Wheaton™ 370120

183.35 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


Product Code. 15699384

  • 198.00€ / Pack of 200
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High temperature autoclave bags manufactured from strong HDPE
  • Suitable for use up to 132°C
  • Uniform wall thickness and seamless bottom, designed to ensure high pressure bearing
  • Printed with blue bio-hazard labelling specifically designed for contaminated waste disposal
  • Write on panel to enable the user to mark each bag as required
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclave Bags
Autoclavable
Up to 132°C
Printed label
HDPE
610 x 810 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers