Learn More
Thermo Scientific™ Rapid Mixing Accessory for Evolution™ Spectrophotometers
Provides reliable and accurate measurement of millisecond timescale kinetics, allowing scientists to make kinetic measurements nearly 1000 times faster than allowed by manual reagent mixing.
21225.00€
Specifications
For Use With (Equipment) Evolution Pro UV-Visible Spectrophotometer
View More Specs
Products 1
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
11883664
View Documents
Thermo Scientific™
222-234400
Pneumatic Drive
21225.00€
Each
Estimated Shipment: 20-02-2025
Description
The Thermo Scientific™ Rapid Mixing Accessory, when used with the Evolution Pro UV-Visible Spectrophotometer, provides reliable and accurate measurement of millisecond timescale kinetics, allowing scientists to make kinetic measurements nearly 1000 times faster than allowed by manual reagent mixing.
Reliable, Millisecond Date Collection Rate
  • Low dead time of 8 milliseconds, when combined with fast data acquisition rate, ensures reliable and precise millisecond data collection
  • 50Hz data collection rate reveals mechanisms hidden by slower measurements
Precise Reaction Start Time
  • Optional automated pneumatic drive mechanism to initiate the reaction and trigger the start of the measurement
  • This air-assisted, mechanical system starts the measurement and eliminates possibility of human error
  • Electronic triggering feature precisely identifies the start of the reaction, enhancing data quality when measuring high performance kinetics
Complete Temperature Control
  • Control and monitor the temperature of the reactants in loading syringes as well as in the reaction component delivery lines supplying the flowcell
  • Includes a thermocouple to measure the temperature of the liquid surrounding the reactants and an alarm to warn if temperature is outside of your specifications
Includes:
Thermocouple and alarm
Compatible with:
  • Evolution Pro UV Visible spectrophotometer
Specifications
Specifications
Evolution Pro UV-Visible Spectrophotometer