Learn More
Rat anti-Mouse IgG2a, Clone: R19-15, biotin, Secondary Antibody, BD
Brand: BD Biosciences 550332
281.83 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The R19-15 antibody recognizes an epitope in the CH3 domain of mouse IgG2a, with strong reactivity to the Igh-I[a] allotype and weaker reactivity to Igh-I[b]. It does not react with other Ig isotypes. Molecular genetic analyses suggest that the Igh-I[b] a
Host Species: Rat
Clone: R19-15
Isotype: IgG1, κ
Species Reactivity: Mouse
Immunogen: Pooled BALB/c and C57BL/6 mouse Ig
ELISA

Specifications
Rat anti-Mouse IgG2a | |
R19-15 | |
Rat | |
RUO | |
Secondary | |
IgG1 κ |
Monoclonal | |
Biotin | |
1 mL | |
Immunology | |
Mouse |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.