Learn More
Rat anti-Mouse IgG2b, Clone: R12-3, biotin, Secondary Antibody, BD
Brand: BD Biosciences 553393
362.02 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The R12-3 antibody recognizes an epitope in the CH3 domain of mouse IgG2b of Igh-C[a] and Igh-C[b] haplotypes. It does not react with other Ig isotypes. Detection of surface immunoglobulin on B lymphoma cells has been demonstrated with R12-3 mAb. This ant
Host Species: Rat
Clone: R12-3
Isotype: IgG2a, κ
Species Reactivity: Mouse
Immunogen: Pooled Mouse Ig
ELISA, Flow Cytometry

Specifications
Rat anti-Mouse IgG2b | |
R12-3 | |
Biotin | |
0.5 mg | |
Immunology | |
Mouse |
Monoclonal | |
0.5mg/mL | |
Rat | |
RUO | |
Secondary | |
IgG2a κ |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.