Learn More
Rat anti-Mouse IgM, Clone: II/41, FITC, Secondary Antibody, BD
Brand: BD Biosciences 553437
263.16 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The II/41 clone has been reported to react specifically with mouse IgM of Igh-C[a] and Igh-C[b] haplotypes. It has been reported not to react with other Ig isotypes. In addition, the II/41 clone has been reported not to stimulate B-cell proliferation. Thi
Host Species: Rat
Clone: II/41
Isotype: IgG2a, κ
Species Reactivity: Mouse
ELISA, Flow Cytometry

Specifications
Rat anti-Mouse IgM | |
II/41 | |
FITC | |
0.5 mg | |
Immunology | |
Mouse |
Monoclonal | |
0.5mg/mL | |
Rat | |
RUO | |
Secondary | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.