missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RBP4/Retinol-Binding Protein 4 Antibody Pair [HRP]
Mouse Monoclonal Antibody Pair
366.00€ - 756.00€
Specifications
Assay Range | 23.44 pg/mL |
---|---|
Assay Sensitivity | 23.44-1500 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Immunoassay Kit Format | Sandwich ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18266567
|
Novus Biologicals™
NBP2-79583-15PLATES |
15 Plates |
756.00€
Pack of 15 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18674921
|
Novus Biologicals™
NBP2-79583-5Plates |
5 Plates |
366.00€
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human RBP4/Retinol-Binding Protein 4.Specifications
23.44 pg/mL | |
Sandwich ELISA | |
Sandwich ELISA Kit | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human RBP4/Retinol-Binding Protein 4. | |
Sandwich ELISA | |
5950 | |
Mouse | |
Cancer, Diabetes Research, Lipid and Metabolism | |
RBP4/Retinol-Binding Protein 4 |
23.44-1500 pg/mL | |
HRP | |
Antibody Pairs | |
Human | |
interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma | |
RBP4 | |
RUO | |
Storage is content dependent. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ RBP4/Retinol-Binding Protein 4 Antibody Pair [HRP]