Learn More
behr Labor-Technik™ Reaction Vessel for COD Determination
Use these vessels for COD Determination. behr Labor-Technik™ Reaction Vessels feature 50 and 100mL scale marks for easy filling prior to titration.
Brand:  behr Labor-Technik™ B00217863
Product Code. 10085100
  • 118.00€ / Each
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 175mL Reaction vessel with vessel adapters
  • NS 29 and 100mL scale mark
  • Flat bottom for improved heat transfer
  • PVDF collar for insulation purposes
  • Gentle transport in EG rack insert
Specifications
Specifications
DIN/DEV
175 mL
Reaction Vessel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
behr Labor-Technik™ Reaction Vessel for COD Determination > Reaction Vessel for COD Determination