missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human HVEM/TNFRSF14 hIgG-His Protein
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
394.00€ - 1390.00€
Specifications
Concentration | 0.25mg/mL |
---|---|
For Use With (Application) | SDS-PAGE |
Formulation | Phosphate buffered saline (pH 7.4) containing 10% glycerol |
Gene ID (Entrez) | 8764 |
Molecular Weight (g/mol) | TMW: 44.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18199489
|
Novus Biologicals™
NBP2-52356-0.01MG |
0.01 mg |
394.00€
0.01mg |
Estimated Shipment: 07-01-2025
Log in to see stock. |
|||||
18232791
|
Novus Biologicals™
NBP2-52356-0.05MG |
0.05 mg |
1390.00€
0.05mg |
Estimated Shipment: 07-01-2025
Log in to see stock. |
|||||
Description
A recombinant protein with a C-Terminal hIgG-His-tag and corresponding to the amino acids 39-202 of Human HVEM/TNFRSF14 The Recombinant Human HVEM/TNFRSF14 Protein is derived from Baculovirus. The Recombinant Human HVEM/TNFRSF14 Protein has been validated for the following applications: SDS-Page.Specifications
0.25mg/mL | |
Phosphate buffered saline (pH 7.4) containing 10% glycerol | |
TMW: 44.6kDa | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
HVEM/TNFRSF14 | |
Recombinant |
SDS-PAGE | |
8764 | |
Baculovirus | |
CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2 | |
Unconjugated | |
>90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |