missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human QPCT Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 6368-ZN-010
This item is not returnable.
View return policy
Description
Measured by its ability to convert Glutaminyl-AMC to pyroglutamyl-AMC. The specific activity is >550 pmol/min/μg, as measured under the described conditions.Specifications
25797 | |
>95% by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
EC 2.3.2.5, GCT, Glutaminyl cyclase, glutaminyl-peptide cyclotransferase, Glutaminyl-tRNA cyclotransferase, Glutamyl cyclase, QCEC, sQC | |
M.W. (Observed): 36-40 kDa, reducing conditions; M.W. (theoretical): 38 kDa | |
10 μg |
Human | |
Q16769 | |
Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol. | |
QPCT | |
Glutaminyl-peptide Cyclotransferase/QPCT | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Glutaminyl-peptide Cyclotransferase/QPCT protein Ala33-Leu361,with an N-terminal 6-His tag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction