missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse LIGHT/TNFSF14 Protein
Recombinant Protein
Brand: Novus Biologicals™ NBP2-61332-100ug
This item is not returnable.
View return policy
Description
LIGHT/TNFSF14 protein is fully biologically active when compared to standard. The ED50 as determined by a cytotoxicity assay using human HT-29 cells is less than 2 μg/mL, corresponding to a specific activity of > 500 IU/mg in the presence of murine anti-polyHistidine monoclonal antibody and rHuIFN-g.Specifications
Western Blot, Bioactivity | |
8740 | |
100 μg | |
Less than 1 EU/μg of LIGHT/TNFSF14 as determined by LAL method. | |
TNFSF14 | |
Apoptosis | |
LIGHT/TNFSF14 |
Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH7.4. | |
MolecularWeight-theroretical: 18.4 kDa | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) | |
CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT | |
Recommended to centrifuge prior to opening. Reconstitute in 100 mM HAC to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) | |
>95%, by SDS-PAGE and HPLC |