Learn More
Kautex™ Red Polypropylene Screw Closures
Seal a container's ingredients with these Kautex™ Red Polypropylene Screw Closures when you package, send samples, transport and store their contents.
Brand: Kautex™ 2000083761
This item is not returnable.
View return policy
Description
- PP
- Red
- Tamper evident ring
- PE-foam insert
Specifications
Red | |
Polypropylene | |
Screw Cap | |
45 mm |
Tamper Evident Ring and Polyethylene Foam Insert | |
Polyethylene Foam Insert | |
100 mL Bottle |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Kautex - Expertise in packaging - Standard plastics
File size: 1.3 MB
Product Content Correction
Product Title
Kautex™ Red Polypropylene Screw Closures > For Use With: 100 mL Bottle