Learn More
Abnova™ RedTaq DNA Polymerase
RedTaq DNA polymerase catalyzes 5'-3' synthesis of DNA.
Brand:  Abnova™ D0012.1000Units
 View more versions of this product
Product Code. 16022555
  • 388.00€ / 1000 units
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The enzyme has been proved not having the 3′-5′ exonuclease activity.
  • The enzyme has proven to have high amplification yield, be stable at high temparetaure.
  • Added inert dye will not have any interference to the reaction.
  • Visual confirmation that the enzyme has been added and proper component mixing of the reaction has occurred.
  • Samples can be loaded directly onto an agarose gel for electrophoresis with no loading dye addition.
Specifications
Specifications
Store at -20°C Guaranteed stable for 6 months when properly stored
Standard PCR
20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol
1000 units
1 unit/μL