Learn More
Witeg™ Reflux Reaction Kit
With a ground neck standard connection of 19/26.
Brand:  Witeg™ A1100200
Product Code. 11847730
  • 162.00€ / Each
Estimated Shipment: 11-04-2025
Description
  • 1 bicolor balloon of 250 mL
  • 1 ball cooler
  • 1 casting bulb of 50 mL
Specifications
Specifications
1 bicolor balloon, 1 casting bulb
1 bulb
ST 19/26
Reflux Reaction Kit
Documents
One moment while we fetch your results.
Product Title
Witeg™ Reflux Reaction Kit > Reflux reaction kit