Learn More
Brady™ Reinforced Polypropylene Mandatory Signs
Brand: Brady™ 252129
This item is not returnable.
View return policy
Specifications
Polypropylene | |
Reinforced Polypropylene | |
120°C | |
Round | |
Mandatory Sign |
1 Ea. | |
200 mm | |
0.369 mm | |
Use Full Protective | |
White on Blue |