missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RelA/NFkB p65 [p Ser276] Inhibitor Peptide Set
Applications: In vivo assay, Functional (Inhibition), Block/Neutralize
636.00€ - 1105.00€
Specifications
Host Species | Human, Mouse, Rat, Bovine, Canine, Mammal |
---|---|
Components | NF-kB p65 (Ser276) Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKQLRRPSDRELSE (p65 sequence: QLRRPSDRELSE). Molecular weight: 3830; Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361 |
For Use With (Application) | Blocking/Neutralizing, Functional (Inhibition), In vivo assay |
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
Product Type | RelA/NFkB p65 [p Ser276] Inhibitor Peptide Set |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18122254
|
Novus Biologicals™
NBP2-26505-5MG |
5 mg |
1105.00€
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18711134
|
Novus Biologicals™
NBP2-26505 |
636.00€
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Specifications
Human, Mouse, Rat, Bovine, Canine, Mammal | |
Blocking/Neutralizing, Functional (Inhibition), In vivo assay | |
RelA/NFkB p65 [p Ser276] Inhibitor Peptide Set | |
Lyophilized |
NF-kB p65 (Ser276) Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKQLRRPSDRELSE (p65 sequence: QLRRPSDRELSE). Molecular weight: 3830; Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
RelA/NFkB p65 [p Ser276] |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ RelA/NFkB p65 [p Ser276] Inhibitor Peptide Set