Learn More
Thermo Scientific™ Cryptococcus Antigen Latex Test Kit
Rapidly detect polysaccharide antigens associated with Cryptococcus neoformans infection using Cryptococcus Antigen Latex Test Kit.
Brand:  Thermo Scientific™ R30851501
582.82 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11974042
  • 605.00€ / 50 tests
Description
Rapidly detect capsular polysaccharide antigens associated with Cryptococcus neoformans infection using Thermo Scientific™ Remel™ Cryptococcus Antigen Latex Test Kit. In 1963, Bloomfield reported detection of cryptococcal polysaccharide (CPS) antigens in serum and CSF of patients with cryptococcosis1. In further testing, investigators reported false-positive reactions in the serum of patients with rheumatoid factor2,3. In 1983, Stockman and Roberts introduced an enzymatic method incorporating protease to eliminate interference factors in serum4.
Cryptococcosis is a systemic infection caused by the yeast, C. neoformans1. The natural reservoirs for C. neoformans are soil and avian feces. Inhalation of yeast cells may lead to a lung infection and possibly disseminated disease. Patients often present with devastating debilitation, especially those with an immunosuppressive syndrome. Because C. neoformans has an unusual affinity for central nervous system tissue, rapid and early detection is essential.
  • Sensitive test
  • Rapid test: Results are obtained within 5 minutes.
  • Easy to identify: A positive test is indicated by any agglutination or clumping and clearing of the test latex immediately after 5 mins rotation step, whereas a negative test is indicated by no agglutination or clumping.
  • Semi-quantitative test: The titer is the reciprocal of the last dilution which produces a positive result (agglutination).
Cryptococcus Antigen Latex Test Kit incorporates the use of latex particles sensitized with murine (mouse) IgM monoclonal antibodies. CPS antigen in patient serum or CSF interacts with sensitized latex particles producing visible agglutination. The use of IgM monoclonal antibodies and treatment of serum with protease reduce the potential for false-positive reactions, eliminating the need to perform a companion control latex test to verify the specificity of results5-6. This test has both diagnostic and prognostic value since progressive disease is usually accompanied by increasing antigen titers. Declining titers are usually associated with clinical improvement, with or without therapy.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cryptococcus Antigen Latex Test Kit
C. neoformans Antigen
1 vial test latex; negative control; positive control; low positive control; protease tablet; 10x specimen diluent; reaction cards; dispensing pipette
50 Tests/Kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P273-Avoid release to the environment.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6196429 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog NumberR30851501
Lot Number6164403 Certificate TypeCertificate of Analysis Date13/12/2024 Catalog NumberR30851501
Lot Number6135896 Certificate TypeCertificate of Analysis Date19/10/2024 Catalog NumberR30851501
Lot Number3787419 Certificate TypeCertificate of Analysis Date14/05/2024 Catalog NumberR30851501
Lot Number3733588 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog NumberR30851501
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Cryptococcus Antigen Latex Test Kit > Latex test; 50/Pk