Learn More
Thermo Scientific™ Remel™ MacConkey Agar
The Thermo Scientific™ Remel MacConkey Agar is used for selective and differential isolation of gram-negative bacilli on the basis of lactose fermentation.
Brand: Thermo Scientific™ R453806
This item is not returnable.
View return policy
Description
Thermo Scientific™ Remel MacConkey Agar is used for selective and differential isolation of gram-negative bacilli on the basis of lactose fermentation.
Specifications
MacConkey Agar | |
Powder | |
Agar | |
10 kg |