Learn More
Thermo Scientific™ Malt Extract Agar (Dehydrated)
Isolate and enumerate yeast and molds from dairy products and other foodstuffs using Malt Extract Agar
Brand: Thermo Scientific™ R453892
187.74 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Isolate and enumerate yeast and molds using Thermo Scientific™ Remel™ Malt Extract Agar (Dehydrated). The utilization of malt or malt extracts for the propagation of yeast and molds is quite common. Reddish described the use of malt extract as a substitute for beer wort in a culture medium1. Thom and Church developed Malt Extract Agar, a modification of Reddish's formula, as a general cultivation medium for aspergilli2. Malt Extract Agar is recommended for the detection of yeasts and molds in dairy products and other foodstuffs3-5.
Isolate and enumerate yeast and molds in dairy products and other foodstuffs using Malt Extract Agar. Both fresh and processed products are subject to fungal spoilage and even heat-processed foods may be affected. Certain foodborne molds and yeasts may also elicit allergic reactions or may cause infections.
This medium contains high concentration of maltose which makes it especially suitable for the growth of yeasts and molds. Dextrin and glycerol are carbon sources and gelatin peptone is a source of nitrogen. The acidic pH of this medium is optimum for the growth of yeasts and molds while inhibiting the growth of bacteria.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
Industrial reference: COMPF. | |
Powder | |
500 g |
Malt Extract Agar | |
Agar |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Not intended for IVD use.