Learn More
Thermo Scientific™ Remel™ McFarland Equivalence Turbidity Standards
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
Brand:  Thermo Scientific™ R20421
340.38 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10036782
  • 372.00€ / 1 set
Description
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
  • Tube size is 15 x 103mm and fits in most spectrophotometers
  • Polystyrene technology allows longer shelf life
Includes:
1 each of 0.5, 1.0, 2.0, 3.0, 4.0.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Turbidity Standard Set (1 each of 0.5, 1.0, 2.0, 3.0, 4.0)
1 each of 0.5, 1.0, 2.0, 3.0, 4.0
15 x 103 mm
Documents
Instructions
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6249825 Certificate TypeCertificate of Analysis Date22/05/2025 Catalog NumberR20421
Lot Number6226137 Certificate TypeCertificate of Analysis Date29/04/2025 Catalog NumberR20421
Lot Number6212738 Certificate TypeCertificate of Analysis Date28/02/2025 Catalog NumberR20421
Lot Number6185616 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberR20421
Lot Number6155589 Certificate TypeCertificate of Analysis Date23/10/2024 Catalog NumberR20421
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Remel™ McFarland Equivalence Turbidity Standards > Turbidity Standard Set