Learn More
Thermo Scientific™ Replacement Seals for Thermo Scientific™ MicroClick and 8 x 8 PCR Strip Rotors
A range of options for secure closure on microcentrifuge rotors to help contain samples.
Brand: Thermo Scientific™ 75005726
126.03 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
A range of options for secure closure on microcentrifuge rotors to help contain samples.
- O-rings for inner and outer seals
- ClickSeal lids for optimal biocontainment
- Available for a variety of sizes and models
Specifications
O-ring | |
MicroClick 30 x 2 and 8 x 8 PCR Strip Rotors |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%