Learn More
Fisherbrand™ Replacement Membrane Kit for Ion Selective Electrode
Allows uptake of the specific ion.
Brand:  Fisherbrand™ I00MK22
 View more versions of this product
Product Code. 11759728
  • 321.18€ / Each
Estimated Shipment: 31-10-2024
Description
  • For Iodide and chloride ISE
Specifications
Specifications
Replacement Membrane Kit
Measuring Iodide ions
0 to 14