Learn More
UVP Replacement Tube
Brand:  UVP 34-0071-01
 View more versions of this product
Product Code. 12882969
  • 61.35€ / Each
Only null left
Estimated Shipment: 10-09-2025
Specifications
UV-B
302 nm
Replacement Tube
Certificates
Promotions
Promotions
Product Content Correction
Product Title
UVP Replacement Tube > Wavelength: 302nm