Learn More
uvex Respirator Face Mask
Breathing protection preformed mask
Brand: uvex 8732310
This item is not returnable.
View return policy
Description
- Preformed mask
- Labelled for use with high levels of dust exposure (D)
Specifications
EN 149, meets the requirements for high dust exposure (D-marking, dolomite dust test), FFP3 protection level | |
Cup/Molded | |
FFP3 | |
Particulate Respirator | |
Yes | |
Classic cup style FFP3 |
White | |
Single-use | |
Polypropylene, Polyester | |
Regular | |
FFP3 | |
15/Pk. |