missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus M2-1 Protein Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93589FR
This item is not returnable.
View return policy
Description
Respiratory Syncytial Virus M2-1 Protein Polyclonal antibody specifically detects Respiratory Syncytial Virus M2-1 Protein in Virus samples. It is validated for Western Blot
Specifications
Respiratory Syncytial Virus M2-1 Protein | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 680 | |
Fusion glycoprotein F0, Fusion glycoprotein F1, Fusion glycoprotein F2, RS virus, RSV | |
Reacts with residues 81-95 [LESYIGSINNITKQSA] of the human Respiratory Syncytial Virus M2 protein, which is a CTL epitope. 93% identity to bovine RSV M2. | |
0.1 mL | |
Virology Bacteria and Parasites | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Respiratory Syncytial Virus M2-1 Protein Antibody, DyLight 680, Novus Biologicals™ > 0.1 mL; DyLight 680
Spot an opportunity for improvement?Share a Content Correction