Learn More
Thermo Scientific™ Resuspension Solution for MagJET Plasmid DNA Kit
The Thermo Scientific MagJET Plasmid DNA Kit is designed for fast and efficient purification of plasmid DNA from overnight cultures of E. coli cultures. The kit utilizes magnetic bead technology, enabling high yields and robust performance.
Brand: Thermo Scientific™ R2582
137.20 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Description
The Thermo Scientific MagJET Plasmid DNA Kit is designed for fast and efficient purification of plasmid DNA from overnight cultures of E. coli cultures. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
Specifications
Suspension Buffer | |
MagJET | |
90 mL |
Kit | |
Resuspension Solution |
For Research Use Only. Not for use in diagnostic procedures.