Learn More
RETSCH ZM 200 Ultra Centrifugal Mill, Push-Fit Rotors
For fine grinding of soft to medium-hard and fibrous materials.
Brand:  RETSCH 02.608.0041
 View more versions of this product
Product Code. 10667931
  • 2225.00€ / Each
Description
  • Accessory for use with ZM 200 Ultra Centrifugal Mill
  • Wear resistant coating
Specifications
Specifications
Stainless Steel
6,000 to 18,000 rpm
Centrifugal Mill
12 Tooth SS - Medium Coarse Materials
9.9 cm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RETSCH ZM 200 Ultra Centrifugal Mill, Push-Fit Rotors > Rotor 12 Tooth S/s Push Lock