Learn More
Thermo Scientific™ Reverse Osmosis Membranes
Choose the required Thermo Scientific™ Reverse Osmosis (RO) Membrane for your Thermo Scientific™ Barnstead™ point of use water purification system, as indicated.
Brand: Thermo Scientific™ 22.0046
147.95 EUR valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Description
Choose the required Thermo Scientific™ Reverse Osmosis (RO) Membrane for your Thermo Scientific™ Barnstead™ point of use water purification system, as indicated.
Use Membranes with the Following Barnstead Systems:
09.2003:
- Reverse osmosis membrane with integrated pretreatment, 3L/hr for Smart2Pure™ systems
09.2006:
- Reverse osmosis membrane with integrated pretreatment, 6L/hr for Smart2Pure systems
22.0046:
- LabTower™ EDI 15 requires two of these RO membranes
- LabTower™ RO 20 requires two of these RO membranes
- LabTower RO 60 requires two of these RO membranes, along with two 22.0087 RO membranes
- LabTower™ TII 20 requires two of these RO membranes
- LabTower TII 60 requires two of these RO membranes, along with two 22.0087 RO membranes
- Pacific™ RO 3, 7, 12 require one of these RO membranes
- Pacific RO 20 requires two of these RO membranes
- Pacific™ TII 3, 7, 12 require one of these RO membranes
- Pacific TII 20 requires two of these RO membranes
22.0087:
- LabTower™ EDI 30 requires two of these RO membranes
- LabTower™ RO 40 requires two of these RO membranes
- LabTower RO 60 requires two of these RO membranes, along with two 22.0046 RO membranes
- LabTower TII 40 requires two of these RO membranes
- LabTower TII 60 requires two of these RO membranes, along with two 22.0046 RO membranes
- Pacific RO 40 requires two of these RO membranes
- Pacific TII 40 requires two of these RO membranes
Specifications
Reverse Osmosis membrane, for use with LabTower EDI 15, LabTower RO 20/60, LabTower TII 20/60, Pacific RO 3/7/12/20, Pacific TII 3/7/12/20 | |
LabTower EDI 15, LabTower RO 20/60, LabTower TII 20/60, Pacific RO 3/7/12/20, Pacific TII 3/7/12/20 | |
Reverse Osmosis Membrane |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59%