Learn More
Grant Instruments™ Reverse Spin Bioreactor for Mixing and Oxygenating Aerobic Cultivation
The Grant Instruments RTS-1 is personal bioreactor which provides 'Reverse-Spin' type of agitation and logging of microbial growth in 50 mL tubes in real time. RTS-1 including TubeSpin™ Bioreactor 50, TPP™, 20 pcs.
Brand: Grant Instruments™ RTS-1 UK
This item is not returnable.
View return policy
Description
Features:- Innovative mixing due to reverse spinning of the sample around its own axis
- Due to innovative mixing technology it is possible to measure optical density of the probe in real time
- Store, display and analyse the data in real time
- Compact device with low profile and small footprint for personal application
- Temperature control allows use as an incubator, e.g. for cell growth
- The ability to change parameters such as temperature, RPM and “Reverse-Spin” frequency, allows to achieve consistency and reproducibility of results
- Programmable Cycling/profiling of cultivation parameters such as temperature, RPM, “Reverse-Spin” frequency
- The ability to remotely monitor the process of cultivation while home or using a mobile phone
Software features:
- Real–Time cell growth logging
- 3D graphical representation of OD or growth rate over time over unit
- Pause option
- Save/Load option
- Report option: PDF and Excel
- Connect up to 12 units simultaneously to 1 computer
- Remote monitoring option (requires internet connection)
- Cycling/Profiling options
- User calibration options for any microorganism
Applications:
- Bacterial cultivation with real time growth kinetics
- Strain screening
- Temperature stress and fluctuation experiments
- Media screening and optimization
- Synthetic and systems biology
- Inhibition and toxicity tests
Specifications
UK Plug |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Grant Instruments™ Reverse Spin Bioreactor for Mixing and Oxygenating Aerobic Cultivation > UK plug