Learn More
Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors
Maintain performance of your Thermo Scientific™ Rheodyne™ Sample Injectors for HPLC with these replacement rotor seals.
130.00€ - 234.32€
Product Code | Brand | For Use With (Equipment) | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Type | Price | Quantity & Availability | ||||
10290285
|
Thermo Scientific™
3725-018 |
Rheodyne Injector Models 3725/3725i/3725-038/3725i-038 | PEEK Replacement Rotor Seals | N/A | |||||
10161263
|
Thermo Scientific™
7010-039 |
Rheodyne Injector Models 7000/7010/7040/7067 | Vespel Replacement Rotor Seals |
192.07€
Each |
Estimated Shipment: 23-01-2025
Log in to see stock. |
||||
10445163
|
Thermo Scientific™
7010-071 |
Rheodyne Injector Models 7000/7010/7040 | ETFE Replacement Rotor Seals |
199.73€
Each |
Estimated Shipment: 13-03-2025
Log in to see stock. |
||||
10671296
|
Thermo Scientific™
7030-003 |
Rheodyne Injector Model 7030 | Vespel Replacement Rotor Seals | N/A | |||||
10576005
|
Thermo Scientific™
7060-070 |
Rheodyne Injector Models 7060/7066 | Vespel Replacement Rotor Seals | N/A | |||||
10648705
|
Thermo Scientific™
7060-074 |
Rheodyne Injector Models 7060/7066/9060 | ETFE Replacement Rotor Seals | N/A | |||||
10150893
|
Thermo Scientific™
7125-047 |
Rheodyne Injector Models 7125/7126 | Vespel Replacement Rotor Seals |
234.32€
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
||||
10269514
|
Thermo Scientific™
7125-079 |
Rheodyne Injector Models 7125/7126 | ETFE Replacement Rotor Seals | N/A | |||||
10427534
|
Thermo Scientific™
8125-038 |
Rheodyne Injector Models 8125/8126 | Vespel Replacement Rotor Seals | N/A | |||||
10510126
|
Thermo Scientific™
8125-097 |
Rheodyne Injector Model 8125 | ETFE Replacement Rotor Seals | N/A | |||||
10179464
|
Thermo Scientific™
9010-051 |
Rheodyne Injector Model 9010 | ETFE Replacement Rotor Seals | N/A | |||||
10280845
|
Thermo Scientific™
9125-082 |
Rheodyne Injector Model 9125 | ETFE Replacement Rotor Seals | N/A | |||||
Description
Use Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors to maintain the performance of your Rheodyne Sample Injectors for high-performance liquid chromatography (HPLC). These seals are suitable for popular Rheodyne injector models and are available in Vespel, PEEK or ETFE.