Learn More
Epredia™ Richard-Allan Scientific™ Eosin-Y Alcoholic, Eosin-Y with Phloxine, Eosin-Y Saturated, Eosin-Y Alcoholic
Enjoy strict quality control of raw materials and finished product and a high quality product every time with the Epredia™ Richard-Allan Scientific™ Eosin-Y Alcoholic, Eosin-Y with Phloxine, and Eosin-Y Saturated.
48.20€ - 161.00€
Product Code | Brand | Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Quantity | Price | Quantity & Availability | ||||
12687806
|
Epredia™
71204 |
Alcoholic Eosin Y Stain | 1 pt. |
53.25€
Pack of 4 |
In Stock
Log in to see stock. |
||||
10046868
|
Epredia™
71211 |
Alcoholic Eosin Y Stain | 1 gal. | N/A | |||||
10249758
|
Epredia™
71304 |
Eosin Phloxine Stain | 1 pt. |
48.20€
Pack of 4 |
In Stock
Log in to see stock. |
||||
Description
- Strict quality control of raw materials and finished product ensures a high quality product every time
- Eosin-Y Alcoholic produces three distinct color variations enhancing differentiation between muscle, red blood cells, and connective tissue
- Eosin-Y w/Phloxine yields a brighter reddish hue and a more dramatic differentiation of cytoplasmic color than Eosin-Y Alcoholic
- Eosin-Y Saturated is a very intense and rapid counterstain that can be used to make working solutions of Eosin-Y
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- EUH210-Safety data sheet available on request.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethyl alcohol, isopropyl,methyl alcohol,acetic acid, acid red 87